Imprimer
Cut resistant gloves, Krytech 579
Fournisseur: MAPA
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
34579088 34579089 34579080 34579081 34579086 34579087
MAPA34579088EA 13.2 EUR
MAPA34579088 MAPA34579089 MAPA34579080 MAPA34579081 MAPA34579086 MAPA34579087
Cut resistant gloves, Krytech 579
Gants
These seamless HDPE liners with PU coating provide standard protection for intricate handling in reasonably clean environments.
  • Suitable for long-term wear: excellent abrasion resistance
  • Excellent dexterity thanks to anatomical shape
  • Ergonomic to reduce fatigue in the hand, with ventilated back
  • Excellent tactile sensitivity due to the gloves reduced thickness
Ideal for precision and automotive assembly, the assembly of thin metal sheets, handling untrimmed parts or sheet metal, handling and sorting small, sharp parts, finishing work as well as for handling cardboard and paper.
Certifications: EN 388:2016 (4342B), ISO 13997: 5.3N (540 g); Cat. II