Imprimer
Chemical resistant gloves, Alto 299
Fournisseur: MAPA
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
34299188 34299189 34299187 34299180
MAPA34299188EA 261 EUR
MAPA34299188 MAPA34299189 MAPA34299187 MAPA34299180
Chemical resistant gloves, Alto 299
Gants
Natural latex gloves with flocked interior.
  • Longer service life and good impact protection, due to the thickness of the material
  • Embossed texture for improved grip
  • Good mechanical performance for long-lasting chemical protection
Ideal for maintenance tasks in in wet environments (contact with water, oil, greases, hydrocarbons) and for emptying and cleaning septic tanks.
Certifications: EN 388:2016 (3121X), EN ISO 374-1 Type A (AKLMPT), EN ISO 374-5; Cat. III