Imprimer
384-well microplates, Falcon®
Fournisseur: Corning
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
353962 353961 353963 353988
736-2044EA 922 EUR
736-2044 736-2043 736-2045 736-2057
384-well microplates, Falcon®
Microplaques Microplaques HTS
Falcon® 384 microplates are compatible with automation systems and meet standard ANSI/SBS footprint dimensions for microplates.