Imprimer
Secure cabinets, filtering ventilation
Fournisseur: TRIONYX
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
AF5-2X AFP5-2X
TRIOAF5-2XEA 2780 EUR
TRIOAF5-2X TRIOAFP5-2X
Secure cabinets, filtering ventilation
Mobilier Armoires de stockage
Secure cabinets are equipped with ventilation system with lighting control. It is an independent cabinet ensuring perfect security and eliminate dangerous vapours.
  • Built in steel 10/10th
  • White epoxy paint RAL 9010
  • Steel containment shelves adjustable in height
  • Removable bottom containment sump
  • Available with polymethylmethacrylate glazed doors or with steel doors
It features with independent storage compartments, self-latching doors with key locks, high and low vents with inlet with fire grid and outlet Ø 100 mm with fire grid.
Specialised high efficiency active charcoal filter and dust pre-filter can be used for treating chemical products.
Certifications: Conforms with NFX15-211 norm, large warning labels in compliance with ISO 3864 and NF X08,003 norms, 92/58/CEE European directive and the health regulation articles.
Informations de commande: Additional shelf, active charcoal filter, containment shelf and base trough in PVC, control of filter saturation level by filter alarm and reactive tubes, industrial absorbent mat for retention tank can be ordered separately. Supplied with noiseless fan (220 to 240 V), cable and plug.