Imprimer
Colonnes capillaires, TraceGOLD™ TG-VMS
Fournisseur: Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
26080-3410 26080-4950 26080-3330 26080-3320
555-8948EA 1690 EUR
555-8948 555-8944 555-8950 555-8951
Colonnes capillaires, TraceGOLD™ TG-VMS
Colonnes pour chromatographie
TraceGOLD™ TG-VMS GC columns are ideal for volatile organic pollutants.
  • Highly stable polymer phase
  • Low bleed for excellent signal-to-noise ratio, sensitivity and mass spectral integrity