Imprimer
Colonnes capillaires, TraceGOLD™ TG-XLBMS
Fournisseur: Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
26079-5780 26079-1540 26079-1300 26079-1430 26079-1550 26079-2230 26079-1420
555-8952EA 843 EUR
555-8952 555-8962 555-8966 555-8963 555-8961 555-8960 555-8964
Colonnes capillaires, TraceGOLD™ TG-XLBMS
Colonnes pour chromatographie
These GC columns are ideal for analysis of active, high molecular weight compounds with sensitive GC-MS systems.
  • Low polarity phase, proprietary
  • Low bleed for excellent signal-to-noise ratio, sensitivity and mass spectral integrity