Imprimer
Magnetic separation devices, Axygen®
Fournisseur: Corning
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
IMAG-12T IMAG-96P
733-2133EA 991 EUR
733-2133 733-2134
Magnetic separation devices, Axygen®
Portoirs Portoirs de séparation magnétique
The magnetic separation devices (MSD) are essential in any paramagnetic bead-based purification process. These IMAG™ devices are designed for a manual process.
  • Single tube or 96-well format
  • Applications include nucleic acid purification, cell based assays and antobody and protein purifications
  • Strong magnets enable separation times of less than 30 seconds
  • Manual operation for either low or high throughput