Imprimer
Container with security sealed screw cap, EUROTUBO®
Fournisseur: DELTALAB
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
444603300 444603402
216-4053EA 535 EUR
216-4053 DELT444603402
Container with security sealed screw cap, EUROTUBO®
Récipients de collecte d'échantillon Conteneurs à usage universel
Containers made of HDPE, translucent, with screw cap, leakproof closure with 'antichildren' system and split seal, ideal for liquids.
  • Good chemical resistance
  • Good temperature resistance
The cap is compound by two pieces that turn around themselves avoiding the cap to be opened, working as a security closure avoiding the accidental manipulation. To open the cap push it down and the sealed caps need to be unscrewed to break the seal, showing that it has not been manipulated.
Attention: *Le(s) produit(s) sur cette page sont hors du champ d'En savoir plus
Commander maintenant
SPÉCIFICATIONS
Environmentally Preferable