Imprimer
Bottles, wide neck, with star-shaped cap
Fournisseur: DELTALAB
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
19411 19408 19409 19410 19412
DELT19411EA 94.5 EUR
DELT19411 DELT19408 DELT19409 DELT19410 DELT19412
Bottles, wide neck, with star-shaped cap
Flacons Flacons en plastique
HDPE, translucent.
  • Tagging points on neck and cap for a tamper proof closure or for labelling
  • Sloping shoulders for easy pouring
  • Moulded-in graduations
Star-shaped screw caps made of polypropylene.
Informations de livraison: Bottles are supplied plugged and capped.