Imprimer
Mixing measuring cylinders, tall form, class A, borosilicate glass
Fournisseur: Kavalier
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
632432630023 632432630025 632432630030 632432630044 632432210923 632432211125 632432211130 632432211238 632432211343 632432211444 632432210718 632432630043
KAVA632432630023EA 248 EUR
KAVA632432630023 KAVA632432630025 KAVA632432630030 KAVA632432630044 KAVA632432210923 KAVA632432211125 KAVA632432211130 KAVA632432211238 KAVA632432211343 KAVA632432211444 KAVA632432210718 KAVA632432630043
Mixing measuring cylinders, tall form, class A, borosilicate glass
Éprouvettes Éprouvettes graduées
Borosilicate glass 3,3, class A.
  • Hexagonal glass base
  • Blue graduations
  • Available with glass or plastic stopper
Certifications: DIN EN ISO 4788