Imprimer
Flasks, pear- shaped, with standard ground joint glass stopper
Fournisseur: Kavalier
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
632426372250 632426372025
KAVA632426372250EA 370 EUR
KAVA632426372250 KAVA632426372025
Flasks, pear- shaped, with standard ground joint glass stopper
Ballons Flacons piriformes
Borosilicate glass 3,3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
The conical geometry makes them ideal for small-scale reactions.