Imprimer
Carboys, narrow neck, LDPE, with stopcock
Fournisseur: SEMADENI
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
1023 1024 1025
SEMA1023EA 252 EUR
SEMA1023 SEMA1024 SEMA1025
Carboys, narrow neck, LDPE, with stopcock
Flacons Bonbonnes
LDPE, translucent.
  • Good chemical resistance
  • Good temperature resistance
  • With stopcock and handle