Imprimer
Chromatography tubing
Fournisseur: Trajan Scientific and Medical
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
062401 062406 062415 062416 0624160 0624173 062418
SGEA062401EA 248 EUR
SGEA062401 SGEA062406 SGEA062415 SGEA062416 SGEA0624160 SGEA0624173 SGEA062418
Chromatography tubing
Tuyaux Tubulures à chromatographie
Stainless steel tubing.
  • Smooth and clean bore
  • Supplied in coils
  • Where tubing O.D. is smaller than ¹/₁₆", ends are bushed to ¹/₁₆" and GC ferrules are supplied
  • 5, 10 and 25 m lengths supplied cleaned and passivated