Imprimer
Dropping bottles, glass, with ground-in stoppers
Fournisseur: Kavalier
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
632414109050 632414109100 632414209050 632414209100
KAVA632414109050EA 1060 EUR
KAVA632414109050 KAVA632414109100 KAVA632414209050 KAVA632414209100
Dropping bottles, glass, with ground-in stoppers
Flacons Flacons compte-gouttes
Glass, clear or brown.
  • Good chemical and physical properties
  • For use in applications that do not involve high thermal stresses
Amber to protect light-sensitive substances.