Imprimer
Reagent bottles, narrow or wide neck, with ground-in glass stopper
Fournisseur: Kavalier
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
N632414102050 632414102100 N632414102250 632414102500 N632414102940 N632414102950 N632414202050 632414202100 N632414202250 632414202500 N632414202950 N6324141045009 N632414104940 N632414104950 632414204500 N632414204940 N632414204950 N632414204109 N218000000108 N218000000111
KAVAN632414102050EA 405 EUR
KAVAN632414102050 KAVA632414102100 KAVAN632414102250 KAVA632414102500 KAVAN632414102940 KAVAN632414102950 KAVAN632414202050 KAVA632414202100 KAVAN632414202250 KAVA632414202500 KAVAN632414202950 KAVAN6324141045009 KAVAN632414104940 KAVAN632414104950 KAVA632414204500 KAVAN632414204940 KAVAN632414204950 KAVAN632414204109 KAVAN218000000108 KAVAN218000000111
Reagent bottles, narrow or wide neck, with ground-in glass stopper
Flacons Flacons en verre
Soda-Lime glass, clear or amber.
  • Good chemical and physical properties
  • For use in applications that are not associated with high thermal loads
  • Stable and robust