Imprimer
Capillary columns, TraceGOLD™ TG-WaxMT Metal
Fournisseur: Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
26M88-1420 26M88-1540
555-9112EA 991 EUR
555-9112 555-9110
Capillary columns, TraceGOLD™ TG-WaxMT Metal
Colonnes pour chromatographie
This low-bleed, stable column produces consistent results.
  • Polar phase, polyethylene glycol
  • Polar-deactivated surface tightly binds polymer for excellent thermal stability
  • Resists oxidative damage, damage from strongly acidic or basic volatiles better than silicone solid phases
  • Equivalent to USP G14, G15, G20 and G39 phases
  • Superior inertness ensures excellent peak shape and sensitivity