Imprimer
Capillary columns, TraceGOLD™ TG-5MT metal
Fournisseur: Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
26M98-0350 26M98-1420 26M98-1540
555-9102EA 573 EUR
555-9102 555-9100 555-9036
Capillary columns, TraceGOLD™ TG-5MT metal
Colonnes pour chromatographie
These columns give greater stability and inertness at elevated temperatures and when working with highly active compounds. They provide greater robustness under increased stress.