Imprimer
Capillary columns, TraceGOLD™ TG-1MT metal
Fournisseur: Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
26M99-1300 26M99-1420
555-9026EA 556 EUR
555-9026 555-9025
Capillary columns, TraceGOLD™ TG-1MT metal
Colonnes pour chromatographie
These non polar, metal columns allow you to work at temperatures up to 430 °C with no compromise in performance when analysing highly active and aggressive compounds.
  • Nonpolar, 100% dimethyl polysiloxane
  • Low bleed for excellent signal-to-noise ratio, sensitivity and mass spectral integrity
  • Stable to 430 °C
These columns are recommended for hydrocarbons, GROs, drugs of abuse and PCBs.