Imprimer
Pipette stand, rotary, for PIPETMAN® M pipettes, Power Carrousel™
Fournisseur: GILSON
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
F807023 FB1001
GILSF807023EA 90.2 EUR
GILSF807023 GILSFB1001
Pipette stand, rotary, for PIPETMAN® M pipettes, Power Carrousel™
Portoirs pour pipettes
This pipette carousel is a comfortable charging solution for up to 5 PIPETMAN® M pipettes that provides optimum protection from shocks and contamination and guarantees continous work.
  • Charges one PIPETMAN® M in only 1 hour (at 80% of its capacity and 100% in 3 hours)
  • Protects valuable instruments from shocks and contamination
  • Pipettes hang vertically to prevent liquids from running inside the shaft
  • Rotating head gives an easy access to the pipettes
  • Slim design for a neat, tidy and organised work bench
  • Unrollable cable will avoid any cable spaghetti issue