Imprimer
Sleeves for ground joints, Azlon®
Fournisseur: SciLabware
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
SWN804 SWN806 SWN810
AZLOSWN804EA 37.5 EUR
AZLOSWN804 AZLOSWN806 AZLOSWN810
Sleeves for ground joints, Azlon®
Manchons et raccords pour joints NS Manchettes pour rodage de raccordement
Tapered sleeves, made of PTFE, for grease free and high vacuum seals using ground glass joints.
  • Chemical resistance
  • Forms a liquid and gas tight seal
  • Self-lubricating and non contaminating
  • Strong construction with knurled shoulder
Working temperature: −100 to +260 °C.
Working vacuum: ≤10 to 3 atmospheres.
Certifications: Manufactured from USP Class VI grade PTFE, suitable for food contact applications in accordance with USA regulations.