Imprimer
Colonnes capillaires, TRACE™ TR-Wax
Fournisseur: Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
260W296P 260W131P 260W142P 260W143P 260W154P 260W155P 260W223P 260W224P 260W225P 260W286P 260W297P 260W298P 260W309P
555-9413EA 823 EUR
555-9413 555-9424 555-9423 555-9422 555-9421 555-9420 555-9419 555-9418 555-9417 555-9414 555-9412 555-9098 555-9097
Colonnes capillaires, TRACE™ TR-Wax
Colonnes pour chromatographie
General-purpose, high polarity columns ideal for esters, alcohols, ketones, glycols and aromatic isomers.
  • Polar phase, polyethylene glycol
  • Highly crosslinked and fully deactivated
  • Solvent washable