Imprimer
Pallets and sumps for IBC storage
Fournisseur: asecos
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
15477 15479 15480
MSGA15477EA 1340 EUR
MSGA15477 MSGA15479 MSGA15480
Pallets and sumps for IBC storage
Stockage des fûts
Blue polyethylene sump pallet with 1100 L capacity for the storage of IBC.
  • Compact and robust design
  • Also available with an integrated pallet shape storage platform made of polyethylene
  • Ideal for storing 1000 litres IBC or small containers
Approved for the safe storage of water pollutants.
Ideal for storing liquids with a maximum specific weight of up to 1800 kg/m³.