Imprimer
Sumps for IBC station with platform
Fournisseur: asecos
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
13054 13055
MSGA13054EA 1430 EUR
MSGA13054 MSGA13055
Sumps for IBC station with platform
Stockage des fûts
Polyethylene sumppallet with 1000 litres capacity for the storage of IBC.
  • Complete corrosion protection
  • Optionally with integrated dispensing area
  • Integrated polyethylene storage platform acts as a safety measure
Compact and robust design. Safe and approved for the safe storage of water pollutants.