Imprimer
Dosing bottles, LDPE
Fournisseur: SEMADENI
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
5471 5469
SEMA5471EA 7.1 EUR
SEMA5471 SEMA5469
Dosing bottles, LDPE
Flacons Flacons compte-gouttes
Wide neck translucent LDPE dosing bottles with white measuring cap and are suitable for viscous liquids.
  • Dose opening of Ø 5 mm
  • Neck Ø 44 mm
  • Good chemical and temperature resistance