Imprimer
Colonnes capillaires, TraceGOLD™ TG-1301MS
Fournisseur: Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
26091-1420 26091-1430 26091-1540 26091-2960 26091-2970 26091-2980 26091-3080 26091-3330 26091-3350 26091-3390 26091-3410 26091-3960 26091-4080
555-8558EA 813 EUR
555-8558 555-8557 555-8555- 555-8728 555-8727 555-8726 555-8725 555-8722 555-8720 555-8717 555-8716 555-8714 555-8713
Colonnes capillaires, TraceGOLD™ TG-1301MS
Colonnes pour chromatographie
TraceGOLD™ TG-1301MS columns have excellent reproducibility and column-to-column consistency even with sensitive detectors like ECDs and MS.
  • Low bleed
  • Low to mid-polarity phase, 6% cyanopropylphenyl/94% dimethyl polysilo
  • Long lifetime
  • Excellent inertness
  • Equivalent to USP G43 phase