Imprimer
Purospher® STAR RP-8 HPLC Columns, Endcapped
Fournisseur: Merck
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
1.50031.0001 1.50032.0001 1.50033.0001 1.50035.0001 1.50038.0001 1.50270.0001 1.50271.0001 1.50272.0001 1.50274.0001 1.51453.0001 1.51454.0001 1.50171.7220 1.51916.0001 1.51917.0001
1.50031.0001EA 724 EUR
1.50031.0001 1.50032.0001 1.50033.0001 1.50035.0001 1.50038.0001 1.50270.0001 1.50271.0001 1.50272.0001 1.50274.0001 1.51453.0001 1.51454.0001 1.50171.7220 1.51916.0001 1.51917.0001
Purospher® STAR RP-8 HPLC Columns, Endcapped
Colonnes pour chromatographie
Purospher® STAR RP-8 endcapped, like Purospher® STAR RP-18 endcapped, is based on high purity silica and has an almost complete surface coverage. Thus, Purospher® STAR RP-8 provides excellent peak symmetry for acidic, basic and even chelating compounds, highest column efficiency in terms of the number of theoretical plates, and exceptional stability from pH 1,5 to 10,5. In addition, they are ideal for a wide range of applications. As the sorbent is less hydrophobic than Purospher® STAR RP-18 endcapped, analytes will typically elute faster on the C8 phase. Purospher® STAR RP-8 endcapped provides enhanced selectivity for positional isomers and symmetrical peak shapes for strongly basic and less hydrophobic compounds.
  • Sélectivité renforcée pour isomères de position
  • Excellente symétrie des pics pour composés polaires et basiques
Les cartouches LiChroCART® s'utilisent avec l'accessoire manu-CART® NT 1.51486.0001, permettant aussi d'insérer une précolonne 4-4 mm. Les LiChroCART® 250×10 mm nécessitent le manu-CART® 10-II 1.51419.0001.
Hibar® RT columns are complete with end fittings. When using a guard column with a Hibar® RT column, 1.51487.0001 guard column cartridge holder for 4×4 mm guard column cartridges LiChroCART® is recommended.