Imprimer
HPLC columns, Purospher® STAR Si and NH₂
Fournisseur: Merck
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
1.50244.0001 1.50245.0001 1.50247.0001 1.50248.0001 1.50249.0001 1.50267.0001 1.50268.0001 1.50269.0001 1.50356.0001 1.50357.0001
1.50244.0001EA 658 EUR
1.50244.0001 1.50245.0001 1.50247.0001 1.50248.0001 1.50249.0001 1.50267.0001 1.50268.0001 1.50269.0001 1.50356.0001 1.50357.0001
HPLC columns, Purospher® STAR Si and NH₂
Chromatography Columns
Purospher® STAR HPLC columns based on highest purity silica are also available for normal phase separations.
Purospher® STAR NH₂ (amino phase) is primarily designed for carbohydrate analysis with a typical mobile phase consisting of acetonitrile and water. Additionally, Purospher® STAR NH₂ can also be used in the normal phase retention mode.
Purospher® STAR NH₂ (amino phase) is primarily designed for carbohydrate analysis with a typical mobile phase consisting of acetonitrile and water. Additionally, Purospher® STAR NH₂ can also be used in the normal phase retention mode.
These LiChroCART® cartridges require manu-CART® cartridge column holder part number 1.51486.000, which can be used to hold one cartridge column with or without a 4×4 mm guard column.