Imprimer
Packaging bottles, narrow neck, rectangular, HDPE, with screw caps
Fournisseur: SEMADENI
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
2026 2025 2024
215-0240EA 293 EUR
215-0240 SEMA2025 SEMA2024
Packaging bottles, narrow neck, rectangular, HDPE, with screw caps
Flacons Flacons en plastique
HDPE, translucent, with thread.
  • Good chemical resistance
  • Good temperature resistance
Informations de commande: Screw caps must be ordered separately.