Imprimer
Sulphur standard dissolved in oil, 1,000 µg/g S in mineral oil Specpure® for wear metal analysis
Fournisseur: Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Specpure®
43869.18EA 115 EUR
43869.18 43869.1Y
Sulphur standard dissolved in oil, 1,000 µg/g S in mineral oil Specpure® for wear metal analysis
Étalons Étalons cationiques Standards de sulfide
Physical form: liquid
MDL No.: MFCD00242296