Imprimer
Zinc standard dissolved in oil, 1,000 µg/g Zn in mineral oil Specpure® for wear metal analysis
Marques : Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
14200.1Y 14200.18
14200.1YEA 0 EUR
14200.1Y 14200.18
Zinc standard dissolved in oil, 1,000 µg/g Zn in mineral oil Specpure® for wear metal analysis
Étalons Étalons cationiques Standards de zinc
Physical Form: Liquid
MDL No.: MFCD00011291