Imprimer
Folic acid, crystallin
Marques : Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Synonyme(s): Vitamin M, (S)-2-(4-(((2-Amino-4-oxo-3,4-dihydropteridin-6-yl)methyl)amino)benzamido)pentanedioic acid, Pteroyl-L-glutamic acid, PteGlu, Vitamin B9
J60833.14 J60833.22
J60833.14EA 100 EUR
J60833.14 J60833.22
Folic acid, crystallin
Folic acid
CAS No.: 59-30-3 Molecular Formula: C19H19N7O6 Formula Weight: 441.41 Physical Form: Crystalline Appearance: Yellow to orange Beilstein Reference: 100781 Merck Reference: 14,4221 MDL No.: MFCD00079305
Vitamin B9. A B-complex vitamin necessary for the body to produce red blood cells
Formule: C₁₉H₁₉N₇O₆
Poids moléculaire: 441.41 g/mol
Température de stockage: Température ambiante
Numéro MDL: MFCD00079305
Numéro CAS: 59-30-3
EINECS: 200-419-0
Indice Merck: 14,4221