N-Bromoacetamide 95%
Marques : Thermo Fisher ScientificTotal Evaluation: 0
Moyenne des Notes : 0.0 sur 5
|
Danger
|
Purity: 95%
CAS No.: 79-15-2
Molecular Formula: C2H4BrNO
Formula Weight: 137.97
Storage Temperature: 2°C to 8°C
Melting Point: 102-106°
Beilstein Reference: 969346
Merck Reference: 14,1398
MDL No.: MFCD00037097
Formule:
C₂H₄BrNO Point de fusion: 102…106 °C |
Numéro MDL:
MFCD00037097 Numéro CAS: 79-15-2 EINECS: 201-181-0 UN: 3261 ADR: 8,III Indice Merck: 14,1398 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...