Imprimer
Funnels with short stem
Fournisseur: SEMADENI
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
0198 0199 0196 0197 0194 0195 0203 0200 0201 0202
221-1141EA 11.5 EUR
221-1141 221-1143 221-1151 221-1152 221-1155 221-1156 SEMA0203 SEMA0200 SEMA0201 SEMA0202
Funnels with short stem
Entonnoirs Entonnoirs pour la filtration Entonnoirs filtrants
Funnels, made of HDPE, with smooth internal walls. Robust and strong. Multipurpose funnel for a wide variety of uses. Suspension eye for easy storage.
  • Easy to clean
  • Good chemical resistance
  • Good temperature resistance