Imprimer
Mounting medium, Entellan® new
Marques : Merck
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
1.07961.0100 1.07961.0500 1.00869.0500
1.07961.0100EA 45.6 EUR
1.07961.0100 1.07961.0500 1.00869.0500
Mounting medium, Entellan® new
Stains Microscopy and staining reagents
Entellan® new is a new mounting medium for cover slippers and is specially suited for commercial automated mounting instruments that operate with cover slippers. Its viscosity ranges between 250 and 600 mPas, 1,00869 has a small viscosity range. This narrow viscosity range improves and accelerates the application, as neither the dropping rate nor the volume need to be reset after the first run with a new bottle of Entellan® new.
  • Easy application
  • Small viscosity range, 500 to 600 mPas (1,00869)
  • Glass bottle