Imprimer
Fernbach culture flasks, with DeLong® style neck
Fournisseur: BELLCO GLASS
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
2550-02800 2554-02800 2554-01800 2554-02833
BELC2550-02800EA 1150 EUR
BELC2550-02800 BELC2554-02800 BELC2554-01800 BELC2554-02833
Fernbach culture flasks, with DeLong® style neck
Ballons Flacons de culture
Manufactured from type 1 class A borosilicate glass, these flasks are designed for culturing organisms requiring a large surface area-to-volume ratio.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Commander maintenant
SPÉCIFICATIONS
Environmentally Preferable