Imprimer
Sampling valves
Fournisseur: Merck
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
MXPESPTCL MXPESP50R MXPESP5CN MXPESP5VT
MILXMXPESPTCLEA 0 EUR
MILXMXPESP14N MILXMXPESP18N MILXMXPESPTCL MILXMXPESP50R MILXMXPESP5CN MILXMXPESP5VT
Sampling valves
Vannes Vannes d'injection
The sampling valves are used for sterile sampling of water or other final products in the pharmaceutical industry.
  • Collect samples mid-stream and filter through a MicropreSure unit, or sample into sterile sampling bags or a graduated cylinder
  • Samples are kept in sanitary condition for immediate sampling with alcohol or hydrogen peroxide injections
Materials: stem and body: 316L stainless steel or ECTFE body in ECTFE-type units, valve cap: ABS or PP, O-ring and plug seal: silicone or fluoroelastomer.
Certifications: Meets the FDA indirect foodEn savoir plus
Commander maintenant
SPÉCIFICATIONS
Environmentally Preferable