Imprimer
VWR®, Faeces/Specimen Containers, Conical, with Screw Cap
Fournisseur: VWR Collection
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
216-0892EA 128 EUR
216-0892 216-0893
VWR®, Faeces/Specimen Containers, Conical, with Screw Cap
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Faeces Sample Collection
PP, translucent.
  • Frosted label, graduated
  • Available with or without spoon
  • Available as sterile or packed under aseptic conditions
Ideal as faeces or general specimen containers.
Cdt: Not individually wrapped.