Imprimer
VWR® Q Path® Safety Pad, Tampons neutraliseurs de formol, tampon de sécurité
Fournisseur: VWR Collection
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
720-2251EA 280 EUR
720-2251 720-2252 720-2253
VWR® Q Path® Safety Pad, Tampons neutraliseurs de formol, tampon de sécurité
Tampons de biopsie
Absorbs the odours (neutralises formalin). Ensures good contrast for biopsies.
  • Different sizes available
  • Very useful for the bottom of transport boxes and small formalin spills
Protects knives and cutting boards against premature wear.