Imprimer
Universal chemical sorbent kits, trolley
Fournisseur: asecos
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
14416
MSGA14416EA 157 EUR
MSGA14416
Universal chemical sorbent kits, trolley
Matériaux absorbants
Mobile emergency spill kit for the safe absorption of acids, alkalis, aggressive chemicals and oil.