Imprimer
Universal socks
Fournisseur: asecos
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
11347 9954
MSGA11347EA 170 EUR
MSGA11347 MSGA9954
Universal socks
Matériaux absorbants
Universal socks for safe absorption of oil, water, cooling liquids and non aggressive chemicals.
  • Optimal for long term use
  • Capacity up to 16 times of its own weight
  • High rib and rub resistance
For containment and absorption of larger leaks.