Imprimer
Universal mats
Fournisseur: asecos
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
11326 9951
MSGA11326EA 196 EUR
MSGA11326 MSGA9951
Universal mats
Matériaux absorbants
Universal mats for safe absorption of oil, water, cooling liquids and non-aggressive chemicals.
  • Capacity up to 16 times of its own weight
  • High rib and rub resistance
  • Mats are pre-cut: only take sufficient material as required