Imprimer
Instrument trays
Fournisseur: VITLAB
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
72298 79890 79990 72398 72498
216-0287EA 40.9 EUR
216-0287 VITL79890 VITL79990 VITL72398 VITL72498
Instrument trays
Plateaux/Récipients/Pots Plateaux pour instruments
Made of melamine, white.
  • Tall shape
  • Rounded corners
  • Smooth surfaces, easy to clean
Practical tray for instruments, tools, and sensitive utensils. Stable and self-supporting.
Informations de commande: Fitting lid made of PS, please order separately.