Imprimer
Analytical funnels, Urbanti Type
Fournisseur: VITLAB
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
325095 325195
VITL325095EA 169 EUR
VITL325095 VITL325195
Analytical funnels, Urbanti Type
Entonnoirs Entonnoirs pour la filtration Entonnoirs analytiques
PMP, crystal clear.
  • Good chemical resistance
  • Temperature resistant up to 175 °C
  • Autoclavable
Urbanti type funnel with spiral ribs for fast filtration. This produces a whirlpool effect so that liquids flow through many times faster. This effect is achieved even with standard filter paper inserted in the funnel. Air is prevented from being trapped between the filter paper and the funnel.