Imprimer
Drum spill trays, PIG®
Fournisseur: NEW PIG
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
DRM369 DRM371
NEWPDRM369EA 131 EUR
NEWPDRM369 NEWPDRM371
Drum spill trays, PIG®
Plateaux/Récipients/Pots Plateaux de déversements
HDPE trays contain messy drips and spills from 210 litre drums.
  • Strong, lightweight polyethylene won't rust or corrode
  • Octagonal shape saves space between drums
  • 210 litre drum in place, trays holds 19 litres
Catch drips, leaks and spills that can make your floor a big mess.