Imprimer
PIG® Absorbent pans
Fournisseur: NEW PIG
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
2410PP 2410PP-01
NEWP2410PPEA 234 EUR
NEWP2410PP NEWP2410PP-01
PIG® Absorbent pans
Matériaux absorbants
Trap annoying drips in this spillproof, absorbent-filled Drip Pan. Polypropylene cover prevents splashes and keeps filler from spilling out.
  • Low-profile drip pan fits underneath drum taps and other tight spaces; wide base won't tip over like unsturdy coffee cans
  • Soft polypropylene cover catches drips without splashing and prevents highly absorbent polypropylene filler from spilling out
  • Highly absorbent polypropylene filler soaks up liquid on contact
  • Ideal for use under leaking drum taps, spigots and flanges to absorb nuisance drips; keeps your floors clean and safe
  • Absorbs and retains oils, coolants, solvents and water
98% pre-consumer recycled polypropylene filler.