Imprimer
Colorants mono-réactifs Amersham Cy3 et Cy5, paquets de 5, Cytiva
Marques : Cytiva
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
PA23031 PA25031
PA23031EA 870 EUR
PA23031 PA25031
Colorants mono-réactifs Amersham Cy3 et Cy5, paquets de 5, Cytiva
Systèmes de liaison et de marquage des protéines
Mono-functional maleimides are particularly suitable for the selective labelling of molecules containing free sulfhydryl groups, such as cysteine residues in proteins and peptides and oligonucleotides.
  • More selective method of antibody labelling that reduces the possibility of the label interfering with subsequent antibody-antigen reactions
  • Provides an alternative when CyDye™ NHS esters are unsuitable
  • Dyes are packaged in premeasured amounts and foil-sealed to ensure consistent labelings
They are also useful in antibody labelling since the dye molecules are introduced away from the antibody binding site, increasing the chance that functionality of the labeled molecule will be unimpaired.
Informations de livraison: Each kit contains five vials of dye, each sufficient for labelling 1 mg of protein.