Imprimer
VWR®, Vacuum Urine Containers
Fournisseur: VWR Collection
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
216-1080EA 167 EUR
216-1080 216-1161 216-2200
VWR®, Vacuum Urine Containers
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Urine Sample Collection/Preparation
The VWR® urine collection container provides a faster and easier alternative for urine collection. This closed system helps prevent pouring off, leakage and wet areas, which positively impacts work environments and conditions.
  • PP, translucent
  • Frosted labelling area
  • Yellow screw cap
Urine collection container with integrated transfer device.