Imprimer
Bottles, water sampling, octogonal, Corning® Gosselin™
Fournisseur: Corning
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
P1000B-51 HP250B-53 P500B-51 P250B-51 HP500B-51 HP500B-50 HP250B-50 HP250B-51
331-0414EA 302 EUR
331-0414 GOSSHP250B-53 GOSSP500B-51 GOSSP250B-51 331-0413 GOSSHP500B-50 GOSSHP250B-50 331-0412
Bottles, water sampling, octogonal, Corning® Gosselin™
Bottles Water Sampling Bottles
The water samling bottles, made of PET, transparent, with screw cap are undosed or dosed with sodium thiosulphate, which inhibits the effects of chlorine, bromine and ozone present in the water when the sample is taken.
  • Molded graduations
  • Unbreakable
  • Tamper-evident screw cap