Imprimer
Shelf pipette holder, model 332
Fournisseur: Socorex Isba
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
320.332.4 320.332
SOCO320.332.4EA 110 EUR
SOCO320.332.4 SOCO320.332
Shelf pipette holder, model 332
Portoirs pour pipettes
Shelf pipette holder made of translucent blue PC. Accommodates all Acura® and Calibra® instruments.
  • Holds two micro, macro and/or multichannel pipettes
  • Self-adhesive strip supplied ensures lasting attachment on any surface