Adenosine-5'-triphosphate disodium salt (ATP disodium salt) hydrate ≥98%
Marques : Thermo Fisher ScientificTotal Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Synonyme(s):
ATP disodium salt hydrate, 5′-ATP-Na2 ATP disodium salt hydrate, Adenosine 5'-Triphosphate Disodium Salt Hydrate, Adenosine-5'-triphosphate disodium salt hydrate, 5'-ATP 2Na Hydrate, ((2R,3S,4R,5R)-5-(6-Amino-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl tetrahydrogen triphosphate disodium salt hydrate, ((2R,3S,4R,5R)-5-(6-Amino-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl tetrahydrogen triphosphate, disodium salt xhydrate, Adenosine 5'-triphosphoric acid disodium hydrate
Purity: 98%
CAS No.: 34369-07-8
Molecular Formula: C10H14N5Na2O13P3
Formula Weight: 551.15
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Sensitivity: Light Sensitive and Hygroscopic
Appearance: White
Merck Reference: 14,155
MDL No.: MFCD00080339
Formule:
C₁₀H₁₄N₅Na₂O₁₃P₃·xH₂O Poids moléculaire: 551.14 g/mol (anhy) |
Numéro MDL:
MFCD00150755 Numéro CAS: 34369-07-8 EINECS: 213-579-1 |